• USA Home
  • 8.41658 - 2,3-Dihydrobenzo(b)furan

EMAIL THIS PAGE TO A FRIEND
8.41658 EMD Millipore

2,3-Dihydrobenzo(b)furan

2,3-Dihydrobenzo(b)furan for synthesis. CAS 496-16-2, molar mass 120.15 g/mol., for synthesis

Synonym: Coumaran

  • CAS Number 496-16-2

  • Linear Formula C₈H₈O

  • Molecular Weight 120.15 g/mol

  •  EC Number 207-817-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8416580005)
bp   188 - 189 °C (1013 hPa)
density   1.082 g/cm3
flash point   66 °C
mp   -21 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.080 - 1.084
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?