• USA Home
  • 8.41663 - Isoquinoline-1-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41663 EMD Millipore

Isoquinoline-1-carboxylic acid

Isoquinoline-1-carboxylic acid for synthesis. CAS 486-73-7, molar mass 173.17 g/mol., for synthesis

  • CAS Number 486-73-7

  • Linear Formula C₁₀H₇NO₂

  • Molecular Weight 173.17 g/mol

  •  EC Number 207-639-6

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   25 g in Plastic bottle (8416630025)
mp   161 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 159 °C
  Melting range (upper value): ≤ 162 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?