EMAIL THIS PAGE TO A FRIEND
8.41667 EMD Millipore

2-Indanone

2-Indanone for synthesis. CAS 615-13-4, molar mass 132.16 g/mol., for synthesis

  • CAS Number 615-13-4

  • Linear Formula C₉H₈O

  • Molecular Weight 132.16 g/mol

  •  EC Number 210-410-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
flash point   100 °C
mp   54 - 56 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 39 00
RTECS 
NK7535500
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?