• USA Home
  • 8.41668 - 2-Methyl-2-pentanol

EMAIL THIS PAGE TO A FRIEND
8.41668 EMD Millipore

2-Methyl-2-pentanol

2-Methyl-2-pentanol for synthesis. CAS 590-36-3, molar mass 102.18 g/mol., for synthesis

  • CAS Number 590-36-3

  • Linear Formula C₆H₁₄O

  • Molecular Weight 102.18 g/mol

  •  EC Number 209-681-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8416680005)
expl. lim.     1.1 - 7 %(V)
vapor pressure   8 hPa (25 °C)
solubility   32.4 g/l
density   0.81 g/cm3 (20 °C)
flash point   21 °C
autoignition temp.   415 °C
mp   -107 °C
shipped in   ambient
storage temp.   Store below +30°C.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 19 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P223: Keep away from any possible contact with water, because of violent reaction and possible flash fire.
P262: Do not get in eyes, on skin, or on clothing.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2560 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2560 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2560 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-37
Flammable.Irritating to respiratory system.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?