• USA Home
  • 8.41670 - 6-Aminopenicillanic acid

EMAIL THIS PAGE TO A FRIEND
8.41670 EMD Millipore

6-Aminopenicillanic acid

6-Aminopenicillanic acid for synthesis. CAS 551-16-6, molar mass 216.26 g/mol., for synthesis

Synonym: 6-APA

  • CAS Number 551-16-6

  • Linear Formula C₈H₁₂N₂O₃S

  • Molecular Weight 216.26 g/mol

  •  EC Number 208-993-4

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds More...
packaging   10 g in Glass bottle (8416700010)
vapor pressure   0.0000038 hPa (25 °C)
solubility   2.46 g/l
mp   205 °C (decomposition)
pH   3.4 (100 g/l, H₂O)
storage temp.   Store at <= 20°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?