• USA Home
  • 8.41675 - 4-Bromo-N,N-dimethylaniline

EMAIL THIS PAGE TO A FRIEND
8.41675 EMD Millipore

4-Bromo-N,N-dimethylaniline

4-Bromo-N,N-dimethylaniline for synthesis. CAS 586-77-6, molar mass 200.07 g/mol., for synthesis

  • CAS Number 586-77-6

  • Linear Formula C₈H₁₀BrN

  • Molecular Weight 200.07 g/mol

  •  EC Number 209-582-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8416750025)
bp   264 °C (1013 hPa)
flash point   113 °C
mp   53 - 55 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 53 °C
  Melting range (upper value): ≤ 55 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?