EMAIL THIS PAGE TO A FRIEND
8.41677 EMD Millipore

2,4-Pentanediol

2,4-Pentanediol for synthesis. CAS 625-69-4, molar mass 104.14 g/mol., for synthesis

  • CAS Number 625-69-4

  • Linear Formula C₅H₁₂O₂

  • Molecular Weight 104.14 g/mol

  •  EC Number 210-907-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8416770025)
bp   114 - 115 °C (33.3 hPa)
density   0.960 g/cm3 (20 °C)
flash point   101 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.958 - 0.962
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 39 95
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?