• USA Home
  • 8.41678 - 3,4-Dimethylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.41678 EMD Millipore

3,4-Dimethylbenzoic acid

3,4-Dimethylbenzoic acid for synthesis. CAS 619-04-5, molar mass 150.17 g/mol., for synthesis

  • CAS Number 619-04-5

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.17 g/mol

  •  EC Number 210-576-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8416780050)
mp   163 - 164 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 163 °C
  Melting range (upper value): ≤ 165 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
RTECS 
DG8734020
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?