• USA Home
  • 8.41679 - L-Methyl phenylalanate hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41679 EMD Millipore

L-Methyl phenylalanate hydrochloride

L-Methyl phenylalanate hydrochloride for synthesis. CAS 7524-50-7, EC Number 231-383-4, chemical formula C₁₀H₁₃NO₂*HCl., for synthesis

  • CAS Number 7524-50-7

  • Linear Formula C₁₀H₁₄ClNO₂

  • Molecular Weight 215.68 g/mol

  •  EC Number 231-383-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8416790005)
mp   152 - 158 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Melting range (lower value): ≥ 152 °C
  Melting range (upper value): ≤ 158 °C
  Spec. rotation [α²0/D (c=2 in ethanol): +37.0 - +39.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?