• USA Home
  • 8.41680 - L-Methyl histidinate dihydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41680 EMD Millipore

L-Methyl histidinate dihydrochloride

L-Methyl histidinate dihydrochloride for synthesis. CAS 7389-87-9, EC Number 230-973-9, chemical formula C₇H₁₁N₃O₂*2HCl., for synthesis

Synonym: L-Histidine methyl ester dihydrochloride

  • CAS Number 7389-87-9

  • Linear Formula C₇H₁₃Cl₂N₃O₂

  • Molecular Weight 242.11 g/mol

  •  EC Number 230-973-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8416800010)
solubility   100 g/l
mp   193 - 200 °C (decomposition)
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Melting range (lower value): ≥ 191 °C
  Melting range (upper value): ≤ 198 °C
  Spec. rotation [α²0/D (c=2 in water): +9.0 - +11.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?