EMAIL THIS PAGE TO A FRIEND
8.41681 EMD Millipore

Dimethyl maleate

Dimethyl maleate for synthesis. CAS 624-48-6, molar mass 144.12 g/mol., for synthesis

  • CAS Number 624-48-6

  • Linear Formula C₆H₈O₄

  • Molecular Weight 144.12 g/mol

  •  EC Number 210-848-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8416810250)
bp   71 °C (1013 hPa)
solubility   80 g/l
density   1.151 g/cm3 (20 °C)
flash point   95 °C
mp   -19 °C
LD 50 oral   LD50 Rat 1410 mg/kg
LD 50 dermal   LD50 Rabbit 610 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 90.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Hazard Statement(s) 
H302: Harmful if swallowed.
H311: Toxic in contact with skin.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?