• USA Home
  • 8.41687 - [2.2]-Paracyclophane

EMAIL THIS PAGE TO A FRIEND
8.41687 EMD Millipore

[2.2]-Paracyclophane

[2.2]-Paracyclophane for synthesis. CAS 1633-22-3, molar mass 208.3 g/mol., for synthesis

  • CAS Number 1633-22-3

  • Linear Formula C₁₆H₁₆

  • Molecular Weight 208.3 g/mol

  •  EC Number 216-644-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8416870001)
mp   282 - 287 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 282 °C
  Melting range (upper value): ≤ 287 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 90 00
RTECS 
YD2404000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?