• USA Home
  • 8.41688 - L-Ethyl alanate hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41688 EMD Millipore

L-Ethyl alanate hydrochloride

L-Ethyl alanate hydrochloride for synthesis. CAS 1115-59-9, EC Number 214-225-9, chemical formula C₅H₁₁NO₂*HCl., for synthesis

Synonym: L-Alanine ethyl ester hydrochloride

  • CAS Number 1115-59-9

  • Linear Formula C₅H₁₂ClNO₂

  • Molecular Weight 153.61 g/mol

  •  EC Number 214-225-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8416880005)
solubility   100 g/l
mp   79 - 81 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Optical rotation α 25/D (c=2.5 in water): +2.4 - +3.4 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?