EMAIL THIS PAGE TO A FRIEND
8.41702 EMD Millipore

4-Nitroimidazole

4-Nitroimidazole for synthesis. CAS 3034-38-6, pH 7 (0.40 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 3034-38-6

  • Linear Formula C₃H₃N₃O₂

  • Molecular Weight 113.08 g/mol

  •  EC Number 221-224-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8417020050)
vapor pressure   1 hPa (20 °C)
solubility   0.40 g/l
autoignition temp.   435 °C
mp   303 °C (decomposition)
pH   7 (0.40 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?