• USA Home
  • 8.41709 - 2-Methylbenzothiazole

EMAIL THIS PAGE TO A FRIEND
8.41709 EMD Millipore

2-Methylbenzothiazole

2-Methylbenzothiazole for synthesis. CAS 120-75-2, molar mass 149.22 g/mol., for synthesis

  • CAS Number 120-75-2

  • Linear Formula C₈H₇NS

  • Molecular Weight 149.22 g/mol

  •  EC Number 204-423-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   237 - 238 °C
density   1.173 g/cm3 (20 °C)
flash point   102 °C
mp   12 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.173 - 1.177
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 20 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?