• USA Home
  • 8.41714 - 3-(Chloromethyl)-pyridinium chloride

EMAIL THIS PAGE TO A FRIEND
8.41714 EMD Millipore

3-(Chloromethyl)-pyridinium chloride

3-(Chloromethyl)-pyridinium chloride for synthesis. CAS 6959-48-4, molar mass 164.03 g/mol., for synthesis

  • CAS Number 6959-48-4

  • Linear Formula C₆H₇Cl₂N

  • Molecular Weight 164.03 g/mol

  •  EC Number 230-150-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8417140005)
mp   143 - 146 °C
LD 50 oral   LD50 Rat 316 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 97.0 %
  Melting range (lower value): ≥ 137 °C
  Melting range (upper value): ≤ 142 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H341: Suspected of causing genetic defects.
Precautionary Statement(s) 
P260: Do not breathe dust.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P281: Use personal protective equipment as required.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
US7000000
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3261 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3261 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3261 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
harmful, corrosive, mutagenic
R Phrase 
R 22-34-68
Harmful if swallowed.Causes burns.Possible risk of irreversible effects.
S Phrase 
S 22-26-36/37/39-45
Do not breathe dust.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?