• USA Home
  • 8.41717 - 2,5-Diphenyl-1,3,4-oxadiazole

EMAIL THIS PAGE TO A FRIEND
8.41717 EMD Millipore

2,5-Diphenyl-1,3,4-oxadiazole

2,5-Diphenyl-1,3,4-oxadiazole for synthesis. CAS 725-12-2, molar mass 222.24 g/mol., for synthesis

  • CAS Number 725-12-2

  • Linear Formula C₁₄H₁₀N₂O

  • Molecular Weight 222.24 g/mol

  •  EC Number 211-968-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   138 - 139 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 138 °C
  Melting range (upper value): ≤ 141 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?