EMAIL THIS PAGE TO A FRIEND
8.41718 EMD Millipore

Formanilide

Formanilide for synthesis. CAS 103-70-8, molar mass 121.13 g/mol., for synthesis

  • CAS Number 103-70-8

  • Linear Formula C₇H₇NO

  • Molecular Weight 121.13 g/mol

  •  EC Number 203-136-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8417180050)
bp   271 °C
solubility   25 - 28 g/l
density   1.132 g/cm3 (50 °C)
mp   46 - 50 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 46 °C
  Melting range (upper value): ≤ 48 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?