EMAIL THIS PAGE TO A FRIEND
8.41719 EMD Millipore

2,4-Dibromoaniline

2,4-Dibromoaniline for synthesis. CAS 615-57-6, molar mass 250.92 g/mol., for synthesis

  • CAS Number 615-57-6

  • Linear Formula C₆H₅Br₂N

  • Molecular Weight 250.92 g/mol

  •  EC Number 210-434-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8417190010)
bp   156 °C (28 hPa)
density   2.26 g/cm3 (20 °C)
mp   80 - 82 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 80 °C
  Melting range (upper value): ≤ 82 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 42 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?