• USA Home
  • 8.41724 - 2-Methylanthraquinone

EMAIL THIS PAGE TO A FRIEND
8.41724 EMD Millipore

2-Methylanthraquinone

2-Methylanthraquinone for synthesis. CAS 84-54-8, molar mass 222.24 g/mol., for synthesis

  • CAS Number 84-54-8

  • Linear Formula C₁₅H₁₀O₂

  • Molecular Weight 222.24 g/mol

  •  EC Number 201-539-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8417240100)
mp   171 - 173 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 167 °C
  Melting range (upper value): ≤ 172 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 69 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?