• USA Home
  • 8.41726 - 5-Chloro-2-pentanone

EMAIL THIS PAGE TO A FRIEND
8.41726 EMD Millipore

5-Chloro-2-pentanone

5-Chloro-2-pentanone for synthesis. CAS 5891-21-4, molar mass 120.57 g/mol., for synthesis

  • CAS Number 5891-21-4

  • Linear Formula C₅H₉ClO

  • Molecular Weight 120.57 g/mol

  •  EC Number 227-565-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417260025)
bp   172 °C (1013 hPa) decomposes
density   1.053 g/cm3 (20 °C)
flash point   73 °C
autoignition temp.   440 °C
LD 50 oral   LD50 Rat 804 mg/kg
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
SA8143500
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?