EMAIL THIS PAGE TO A FRIEND
8.41729 EMD Millipore

8-Bromoadenosine

8-Bromoadenosine for synthesis. CAS 2946-39-6, molar mass 346.13 g/mol., for synthesis

  • CAS Number 2946-39-6

  • Linear Formula C₁₀H₁₂BrN₅O₄

  • Molecular Weight 346.13 g/mol

  •  EC Number 220-959-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8417290001)
mp   211 °C (decomposition)
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Spec. rotation [α²0/D (C=1.07 in 1N HCL): -35.0 - -30.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?