• USA Home
  • 8.41742 - 2-Phenylethyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
8.41742 EMD Millipore

2-Phenylethyl isothiocyanate

for synthesis

  • CAS Number 2257-09-2

  • Linear Formula C₉H₉NS

  • Molecular Weight 163.24 g/mol

  •  EC Number 218-855-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8417420005)
bp   139 - 140 °C (14.7 hPa)
density   1.09 g/cm3 (20 °C)
flash point   100 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.093 - 1.097
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?