EMAIL THIS PAGE TO A FRIEND
8.41743 EMD Millipore

5-Bromouracil

5-Bromouracil for synthesis. CAS 51-20-7, molar mass 190.98 g/mol., for synthesis

  • CAS Number 51-20-7

  • Linear Formula C₄H₃BrN₂O₂

  • Molecular Weight 190.98 g/mol

  •  EC Number 200-084-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8417430025)
solubility   <1 g/l
mp   310 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?