EMAIL THIS PAGE TO A FRIEND
8.41748 EMD Millipore

Azelainitrile

Azelainitrile for synthesis. CAS No. 1675-69-0, EC Number 216-824-0., for synthesis

  • CAS Number 1675-69-0

  • Linear Formula C₉H₁₄N₂

  • Molecular Weight 150.22 g/mol

  •  EC Number 216-824-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417480025)
bp   175 - 176 °C (14.7 hPa)
density   0.923 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.923 - 0.927
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302 + H332: Harmful if swallowed or if inhaled
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 20/22
Harmful by inhalation and if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?