• USA Home
  • 8.41750 - Sodium bis(trimethylsilyl)-amide

EMAIL THIS PAGE TO A FRIEND
8.41750 EMD Millipore

Sodium bis(trimethylsilyl)-amide

(1 M solution in tetrahydrofuran) for synthesis

Purchase

Properties

packaging   100 ml in Glass bottle (8417500100)
expl. lim.     1.5 - 12.4 %(V) (THF)
density   0.9 g/cm3 (20 °C)
flash point   -20 °C (THF)
autoignition temp.   215 °C (THF)
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): 0.90 - 1.10 mol/l

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
3824 99 92
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
H351: Suspected of causing cancer.
EUH019: May form explosive peroxides.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P240: Ground/bond container and receiving equipment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
26
Alkali and alkaline-earth metals should be taken up in an inert solvent and neutralized by adding 2-propanol (Cat. No. 100995) dropwise with stirring. Should the reaction be more violent than expected, conversion should be carried out with tert-butanol or octanol. Caution: This reaction produces hydrogen, which can form explosive mixtures; take necessary precautions. When the reaction has ceased, add water dropwise; neutralize; container D. In the case of alkali or alkaline-earth borohydrides, add methanol (Cat. No. 106008) with stirring; in the case of alkali or alkaline-earth amides and hydrides, organoaluminium and organotin hydrides, add 2-propanol (Cat. No. 100995) dropwise. The substances, which are usually in solid form, should be previously suspended in an ether! When the respective reaction has ceased, hydrolize with water; then neutralize. Container D or E. Lithium aluminium hydride must also be destroyed by slurrying in an ether. Under an inert gas and with thorough stirring, add dropwise a 1:4 mixture of ethyl acetate (Cat. No. 822277) and the ether used to prepare the slurry. Always ensure thorough mixing (stirring)! Container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 2924 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2924 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2924 , 3 (8), II, Segregation Group: 18 (Alkalis)
Categories of danger 
highly flammable, corrosive, carcinogenic
R Phrase 
R 11-19-34-37-40
Highly flammable.May form explosive peroxides.Causes burns.Irritating to respiratory system.Limited evidence of a carcinogenic effect.
S Phrase 
S 16-26-36/37/39-45
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?