EMAIL THIS PAGE TO A FRIEND
8.41760 EMD Millipore

2-Acetylthiazole

2-Acetylthiazole for synthesis. CAS 24295-03-2, molar mass 127.16 g/mol., for synthesis

  • CAS Number 24295-03-2

  • Linear Formula C₅H₅NOS

  • Molecular Weight 127.16 g/mol

  •  EC Number 246-134-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8417600001)
bp   89 - 91 °C (16 hPa)
density   1.227 g/cm3
flash point   78 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.224 - 1.228
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?