• USA Home
  • 8.41761 - trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene

EMAIL THIS PAGE TO A FRIEND
8.41761 EMD Millipore

trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene

trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene for synthesis. CAS 54125-02-9, molar mass 172.3 g/mol. MSDS and product information below., for synthesis

  • CAS Number 54125-02-9

  • Linear Formula C₈H₁₆O₂Si

  • Molecular Weight 172.3 g/mol

Purchase

Properties

bp   68 - 69 °C (18.7 hPa)
density   0.902 g/cm3 (20 °C)
flash point   59 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.900 - 0.904
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, I
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, I
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, I

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?