• USA Home
  • 8.41763 - Ethylcyclopentanone-2-carboxylate

EMAIL THIS PAGE TO A FRIEND
8.41763 EMD Millipore

Ethylcyclopentanone-2-carboxylate

Ethylcyclopentanone-2-carboxylate for synthesis. CAS 611-10-9, molar mass 156.18 g/mol., for synthesis

  • CAS Number 611-10-9

  • Linear Formula C₈H₁₂O₃

  • Molecular Weight 156.18 g/mol

  •  EC Number 210-253-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417630025)
bp   102 - 104 °C (14.7 hPa)
density   1.080 g/cm3 (20 °C)
flash point   77 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.078 - 1.082
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?