• USA Home
  • 8.41767 - 3-Hydroxy-4-methoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.41767 EMD Millipore

3-Hydroxy-4-methoxybenzaldehyde

3-Hydroxy-4-methoxybenzaldehyde for synthesis. CAS 621-59-0, molar mass 152.15 g/mol., for synthesis

  • CAS Number 621-59-0

  • Linear Formula C₈H₈O₃

  • Molecular Weight 152.15 g/mol

  •  EC Number 210-694-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8417670010)
bp   179 °C (20 hPa)
flash point   >100 °C
mp   113 - 116 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 113 °C
  Melting range (upper value): ≤ 116 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 49 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?