EMAIL THIS PAGE TO A FRIEND
8.41769 EMD Millipore

1,5-Hexadiene

1,5-Hexadiene for synthesis. CAS 592-42-7, molar mass 82.14 g/mol., for synthesis

  • CAS Number 592-42-7

  • Linear Formula C₆H₁₀

  • Molecular Weight 82.14 g/mol

  •  EC Number 209-754-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8417690050)
bp   60 °C
expl. lim.     1.2 - 6.1 %(V)
vapor pressure   295 hPa (25 °C)
solubility   169 mg/l
density   0.69 g/cm3 (20 °C)
flash point   -27 °C
mp   -141 °C
refractive index   1.404 (20 °C, 589 nm)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.690 - 0.693
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2901 29 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P223: Keep away from any possible contact with water, because of violent reaction and possible flash fire.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2458 , 3, II
Declaration (transport by air) IATA-DGR 
UN 2458 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 2458 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-36/37
Highly flammable.Irritating to eyes and respiratory system.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?