• USA Home
  • 8.41771 - 2-Aminobenzophenone

EMAIL THIS PAGE TO A FRIEND
8.41771 EMD Millipore

2-Aminobenzophenone

2-Aminobenzophenone for synthesis. CAS 2835-77-0, molar mass 197.23 g/mol., for synthesis

  • CAS Number 2835-77-0

  • Linear Formula C₁₃H₁₁NO

  • Molecular Weight 197.23 g/mol

  •  EC Number 220-613-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8417710010)
bp   280 °C (1013 hPa)
flash point   240 °C
mp   105 - 108 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 105 °C
  Melting range (upper value): ≤ 108 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?