• USA Home
  • 8.41775 - Cyclopropanecarbonitrile

EMAIL THIS PAGE TO A FRIEND
8.41775 EMD Millipore

Cyclopropanecarbonitrile

Cyclopropanecarbonitrile for synthesis. CAS 5500-21-0, molar mass 67.09 g/mol., for synthesis

  • CAS Number 5500-21-0

  • Linear Formula C₄H₅N

  • Molecular Weight 67.09 g/mol

  •  EC Number 226-836-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417750025)
bp   133 - 135 °C (1013 hPa)
density   0.895 g/cm3 (20 °C)
flash point   32 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.893 - 0.897
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 1992 , 3 (6.1), III
Declaration (transport by air) IATA-DGR 
UN 1992 , 3 (6.1), III
Declaration (transport by sea) IMDG-Code 
UN 1992 , 3 (6.1), III
Categories of danger 
flammable, toxic
R Phrase 
R 10-23/24/25
Flammable.Toxic by inhalation, in contact with skin and if swallowed.
S Phrase 
S 36/37-45
Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?