EMAIL THIS PAGE TO A FRIEND
8.41777 EMD Millipore

Pyridazine

Pyridazine for synthesis. CAS 289-80-5, molar mass 80.08 g/mol., for synthesis

  • CAS Number 289-80-5

  • Linear Formula C₄H₄N₂

  • Molecular Weight 80.08 g/mol

  •  EC Number 206-025-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8417770005)
bp   208 °C (1013 hPa)
density   1.106 g/cm3
flash point   85 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.104 - 1.108
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?