• USA Home
  • 8.41779 - 3-Phenoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.41779 EMD Millipore

3-Phenoxybenzyl alcohol

3-Phenoxybenzyl alcohol for synthesis. CAS 13826-35-2, molar mass 200.24 g/mol., for synthesis

  • CAS Number 13826-35-2

  • Linear Formula C₁₃H₁₂O₂

  • Molecular Weight 200.24 g/mol

  •  EC Number 237-525-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8417790010)
bp   135 - 140 °C (13.3 hPa)
vapor pressure   0.13 hPa (86 °C)
density   1.146 g/cm3 (20 °C)
flash point   309 °C
LD 50 oral   LD50 Rat 1496 mg/kg
LD 50 dermal   LD50 Rabbit 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.146 - 1.150
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?