• USA Home
  • 8.41780 - Phenylpyruvic acid sodium salt monohydrate

EMAIL THIS PAGE TO A FRIEND
8.41780 EMD Millipore

Phenylpyruvic acid sodium salt monohydrate

Phenylpyruvic acid sodium salt monohydrate for synthesis. CAS 122049-54-1, molar mass 204.16 g/mol., for synthesis

  • CAS Number 122049-54-1

  • Linear Formula C₉H₇NaO₃ * H₂O

  • Molecular Weight 204.16 g/mol

  •  EC Number 204-053-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8417800005)
mp   260 °C
storage temp.   Store at <= 20°C.
specifications   Assay (HClO₄): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?