• USA Home
  • 8.41781 - 1-Methyl-1-cyclopentene

EMAIL THIS PAGE TO A FRIEND
8.41781 EMD Millipore

1-Methyl-1-cyclopentene

1-Methyl-1-cyclopentene for synthesis. CAS 693-89-0, molar mass 82.15 g/mol., for synthesis

  • CAS Number 693-89-0

  • Linear Formula C₆H₁₀

  • Molecular Weight 82.15 g/mol

  •  EC Number 211-762-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   72 °C (1005 hPa)
density   0.78 g/cm3 (20 °C)
flash point   -17 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.779 - 0.783
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 19 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?