EMAIL THIS PAGE TO A FRIEND
8.41782 EMD Millipore

4-Pentene-2-ol

4-Pentene-2-ol for synthesis. CAS 625-31-0, molar mass 86.13 g/mol., for synthesis

  • CAS Number 625-31-0

  • Linear Formula C₅H₁₀O

  • Molecular Weight 86.13 g/mol

  •  EC Number 210-887-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8417820005)
density   0.837 g/cm3 (20 °C)
flash point   30 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.836 - 0.840
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 29 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1987 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1987 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1987 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?