• USA Home
  • 8.41786 - 3-(Dimethylamino)-phenol

EMAIL THIS PAGE TO A FRIEND
8.41786 EMD Millipore

3-(Dimethylamino)-phenol

3-(Dimethylamino)-phenol for synthesis. CAS 99-07-0, pH 7 (10 g/l, H₂O, 20 °C) (slurry)., for synthesis

  • CAS Number 99-07-0

  • Linear Formula C₈H₁₁NO

  • Molecular Weight 137.18 g/mol

  •  EC Number 202-727-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8417860025)
bp   140 °C (12 hPa)
flash point   141 °C
mp   81 - 84 °C
pH   7 (10 g/l, H₂O, 20 °C) (slurry)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 81 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 21/22-36/37/38
Harmful in contact with skin and if swallowed.Irritating to eyes, respiratory system and skin.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?