• USA Home
  • 8.41791 - N-Methylethylenediamine

EMAIL THIS PAGE TO A FRIEND
8.41791 EMD Millipore

N-Methylethylenediamine

N-Methylethylenediamine for synthesis. CAS 109-81-9, molar mass 74.13 g/mol., for synthesis

  • CAS Number 109-81-9

  • Linear Formula C₃H₁₀N₂

  • Molecular Weight 74.13 g/mol

  •  EC Number 203-707-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8417910005)
bp   114 - 116 °C (1013 hPa)
density   0.841 g/cm3 (20 °C)
flash point   41 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2734 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2734 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2734 , 8 (3), II, Segregation Group: 18 (Alkalis)
Categories of danger 
flammable, corrosive
R Phrase 
R 10-34
Flammable.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?