• USA Home
  • 8.41792 - N,N'-Dimethylethylenediamine

EMAIL THIS PAGE TO A FRIEND
8.41792 EMD Millipore

N,N'-Dimethylethylenediamine

N,N'-Dimethylethylenediamine for synthesis. CAS 110-70-3, molar mass 88.15 g/mol., for synthesis

  • CAS Number 110-70-3

  • Linear Formula C₄H₁₂N₂

  • Molecular Weight 88.15 g/mol

  •  EC Number 203-793-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417920025)
bp   119 °C (1013 hPa)
density   0.82 g/cm3 (20 °C)
flash point   26 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 78.5 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
KV4250000
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2734 , 8 (3), II
Declaration (transport by air) IATA-DGR 
UN 2734 , 8 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2734 , 8 (3), II, Segregation Group: 18 (Alkalis)
Categories of danger 
flammable, corrosive
R Phrase 
R 10-34
Flammable.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?