• USA Home
  • 8.41793 - Glycinamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.41793 EMD Millipore

Glycinamide hydrochloride

Glycinamide hydrochloride for synthesis. CAS 1668-10-6, EC Number 216-789-1, chemical formula C₂H₆N₂O*HCl., for synthesis

  • CAS Number 1668-10-6

  • Linear Formula C₂H₇ClN₂O

  • Molecular Weight 110.54 g/mol

  •  EC Number 216-789-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8417930025)
mp   204 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 201 °C
  Melting range (upper value): ≤ 204 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?