• USA Home
  • 8.41794 - 2,3,5,6-Tetramethylpyrazine

EMAIL THIS PAGE TO A FRIEND
8.41794 EMD Millipore

2,3,5,6-Tetramethylpyrazine

2,3,5,6-Tetramethylpyrazine for synthesis. CAS 1124-11-4, molar mass 136.19 g/mol., for synthesis

  • CAS Number 1124-11-4

  • Linear Formula C₈H₁₂N₂

  • Molecular Weight 136.19 g/mol

  •  EC Number 214-391-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8417940025)
bp   190 °C (1013 hPa)
vapor pressure   70 hPa (50 °C)
solubility   4 g/l
density   1.08 g/cm3 (20 °C)
mp   83 - 85 °C
LD 50 oral   LD50 Rat 1910 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?