• USA Home
  • 8.41795 - 3'-Chloroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.41795 EMD Millipore

3'-Chloroacetophenone

3'-Chloroacetophenone for synthesis. CAS 99-02-5, molar mass 154.59 g/mol., for synthesis

  • CAS Number 99-02-5

  • Linear Formula C₈H₇ClO

  • Molecular Weight 154.59 g/mol

  •  EC Number 202-721-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8417950025)
bp   227 - 229 °C (1013 hPa)
density   1.194 g/cm3 (20 °C)
flash point   105 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.192 - 1.196
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3416 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3416 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3416 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?