EMAIL THIS PAGE TO A FRIEND
8.41801 EMD Millipore

N-Phenylglycin

N-Phenylglycin for synthesis. CAS 103-01-5, molar mass 151.16 g/mol., for synthesis

  • CAS Number 103-01-5

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 203-070-2

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8418010025)
mp   123.5 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 93.0 % (a/a)
  Melting range (lower value): ≥ 120 °C
  Melting range (upper value): ≤ 125 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?