• USA Home
  • 8.41806 - 5-Methyl-2-nitrophenol

EMAIL THIS PAGE TO A FRIEND
8.41806 EMD Millipore

5-Methyl-2-nitrophenol

5-Methyl-2-nitrophenol for synthesis. CAS 700-38-9, molar mass 153.14 g/mol., for synthesis

  • CAS Number 700-38-9

  • Linear Formula C₇H₇NO₃

  • Molecular Weight 153.14 g/mol

  •  EC Number 211-843-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8418060025)
flash point   109 °C
mp   53 - 56 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 52 °C
  Melting range (upper value): ≤ 55 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 99 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
SH1130950
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2446 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2446 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2446 , 6.1, III
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-28-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.After contact with skin, wash immediately with plenty of water.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?