• USA Home
  • 8.41812 - 4-Chloro-3,5-dinitrobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.41812 EMD Millipore

4-Chloro-3,5-dinitrobenzotrifluoride

4-Chloro-3,5-dinitrobenzotrifluoride for synthesis. CAS 393-75-9, molar mass 270.55 g/mol., for synthesis

  • CAS Number 393-75-9

  • Linear Formula C₇H₂ClF₃N₂O₄

  • Molecular Weight 270.55 g/mol

  •  EC Number 206-889-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8418120005)
vapor pressure   0.46 hPa (20 °C)
solubility   0.05 g/l
density   1.6 g/cm3
flash point   126 °C
autoignition temp.   >560 °C
mp   55 - 58 °C
LD 50 oral   LD50 Rat 930 mg/kg
LD 50 dermal   LD50 Rabbit 161 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 55 °C
  Melting range (upper value): ≤ 58 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 99 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H310: Fatal in contact with skin.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
XS9065000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
toxic, irritant
R Phrase 
R 22-24-36/38
Harmful if swallowed.Toxic in contact with skin.Irritating to eyes and skin.
S Phrase 
S 36/37-45
Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?