• USA Home
  • 8.41816 - Biphenyl-4-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.41816 EMD Millipore

Biphenyl-4-carboxylic acid

Biphenyl-4-carboxylic acid for synthesis. CAS 92-92-2, molar mass 198.22 g/mol., for synthesis

  • CAS Number 92-92-2

  • Linear Formula C₁₃H₁₀O₂

  • Molecular Weight 198.22 g/mol

  •  EC Number 202-203-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8418160025)
solubility   0.03 g/l
mp   223 - 228 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 223 °C
  Melting range (upper value): ≤ 228 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
DV1925100
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36
Irritating to eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?