• USA Home
  • 8.41825 - 1,2-Cyclohexanedione dioxime

EMAIL THIS PAGE TO A FRIEND
8.41825 EMD Millipore

1,2-Cyclohexanedione dioxime

1,2-Cyclohexanedione dioxime for synthesis. CAS 492-99-9, molar mass 142.15 g/mol., for synthesis

Synonym: Nioxime

  • CAS Number 492-99-9

  • Linear Formula C₆H₁₀N₂O₂

  • Molecular Weight 142.15 g/mol

  •  EC Number 207-769-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8418250005)
mp   183.8 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?