• USA Home
  • 8.41827 - 2,6-Dichlorotoluene

EMAIL THIS PAGE TO A FRIEND
8.41827 EMD Millipore

2,6-Dichlorotoluene

2,6-Dichlorotoluene for synthesis. CAS 118-69-4, molar mass 161.03 g/mol., for synthesis

  • CAS Number 118-69-4

  • Linear Formula C₇H₆Cl₂

  • Molecular Weight 161.03 g/mol

  •  EC Number 204-269-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8418270100)
bp   196 - 203 °C (1013 hPa)
vapor pressure   34 Pa (25 °C)
solubility   26 mg/l insoluble
density   1.267 g/cm3 (20 °C)
flash point   82 °C
mp   2.8 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.264 - 1.268
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
dangerous for the environment
R Phrase 
R 51/53
Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-61
Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?